Learn More
AdipoGen Butyrolactone 3
SDP
Catalog No. 502562336
Encompass_Preferred
Chemical. CAS 778649-18-6. Formula C9H12O4. MW 184.2. Synthetic Specific histone acetyltransferase Gcn5 inhibitor. Anticancer compound. Pre-mRNA splicing inhibitor. Blocks splicing before the first catalytic steps. Potential anti-inflammatory compound.
Catalog No. 50-256-2336 Supplier AdipoGen Supplier No. AGCR13522M002
Chemical. CAS 778649-18-6. Formula C9H12O4. MW 184.2. Synthetic Specific histone acetyltransferase Gcn5 inhibitor. Anticancer compound. Pre-mRNA splicing inhibitor. Blocks splicing before the first catalytic steps. Potential anti-inflammatory compound.