Learn More
Apexbio Technology LLC Butyrolactone 3 778649-18-6 5mg
SDP
Catalog No. 5000087986
Encompass_Preferred
Butyrolactone 3 (CAS 778649-18-6) is a small-molecule inhibitor that targets the histone acetyltransferase Gcn5 a member of the GNAT family with specificity for histone H3 acetylation In vitro studies demonstrate that Butyrolactone 3 inhibits Gcn5 activity with an IC50 of 100 M while showing minimal inhibition of CBP The binding affinity of Butyrolactone 3 to Gcn5 is comparable to that of histone H3 and its inhibition is non-irreversible indicating it does not act via a Michael addition mechanism Structurally modification at position 2 has been suggested as a basis for structure activity relationship (SAR) studies Currently Butyrolactone 3 is utilized in preclinical research aimed at modulating Gcn5-mediated histone acetylation
Catalog No. 50-000-87986 Supplier Apexbio Technology LLC Supplier No. C32095
Butyrolactone 3 (CAS 778649-18-6) is a small-molecule inhibitor that targets the histone acetyltransferase Gcn5 a member of the GNAT family with specificity for histone H3 acetylation In vitro studies demonstrate that Butyrolactone 3 inhibits Gcn5 activity with an IC50 of 100 M while showing minimal inhibition of CBP The binding affinity of Butyrolactone 3 to Gcn5 is comparable to that of histone H3 and its inhibition is non-irreversible indicating it does not act via a Michael addition mechanism Structurally modification at position 2 has been suggested as a basis for structure activity relationship (SAR) studies Currently Butyrolactone 3 is utilized in preclinical research aimed at modulating Gcn5-mediated histone acetylation