Learn More
Medchemexpress LLC Butyrolactone 3 | 778649-18-6 | 99.6% | 50 MG
SDP
Catalog No. 5000339511
Encompass_Preferred
Butyrolactone 3 (MB-3) is a specific small-molecule inhibitor of the histone acetyltransferase Gcn5, with an IC50 of 100 μM. It exhibits high affinity for the Gcn5 enzyme, similar to its natural substrate, histone H3, and shows weak inhibitory effects on CBP (IC50=0.5 mM). This compound is intended for research use only and is utilized in studies related to cancer, metabolic, autoimmune, and neurological diseases.
  • Specific small-molecule inhibitor of histone acetyltransferase Gcn5
  • Exhibits high affinity for Gcn5 enzyme
  • Shows weak inhibitory effects on CBP
  • Suitable for research in cancer, metabolic, autoimmune, and neurological diseases
Catalog No. 50-003-39511 Supplier Medchemexpress LLC Supplier No. HY12903950MG
Butyrolactone 3 (MB-3) is a specific small-molecule inhibitor of the histone acetyltransferase Gcn5, with an IC50 of 100 μM. It exhibits high affinity for the Gcn5 enzyme, similar to its natural substrate, histone H3, and shows weak inhibitory effects on CBP (IC50=0.5 mM). This compound is intended for research use only and is utilized in studies related to cancer, metabolic, autoimmune, and neurological diseases.
  • Specific small-molecule inhibitor of histone acetyltransferase Gcn5
  • Exhibits high affinity for Gcn5 enzyme
  • Shows weak inhibitory effects on CBP
  • Suitable for research in cancer, metabolic, autoimmune, and neurological diseases