Learn More
Selleck Chemical LLC Butyrolactone 3-E1387-1MG
SDP
Catalog No. 503134892
Encompass_Preferred
Butyrolactone 3 (MB-3) is a specifical small-molecule inhibitor of the histone acetyltransferase Gcn5 (IC50 100 M) which has a high affinity to the Gcn5 enzyme comparable to that of its natural substrate histone H3 Butyrolactone 3 shows weak inhibitory on CBP (IC50 0 5 mM) Butyrolactone 3 can be used in studies of cancer metabolic autoimmune and neurological diseases
Catalog No. 50-313-4892 Supplier Selleck Chemical LLC Supplier No. E13871MG
Butyrolactone 3 (MB-3) is a specifical small-molecule inhibitor of the histone acetyltransferase Gcn5 (IC50 100 M) which has a high affinity to the Gcn5 enzyme comparable to that of its natural substrate histone H3 Butyrolactone 3 shows weak inhibitory on CBP (IC50 0 5 mM) Butyrolactone 3 can be used in studies of cancer metabolic autoimmune and neurological diseases