Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyrolactone 3 (MB-3) | 778649-18-6 | 99.6% | 1 MG
SDP

Catalog No. 5000340480
Encompass_Preferred

Butyrolactone 3 (MB-3) is a specific small-molecule inhibitor of the histone acetyltransferase Gcn5 (IC50=100 μM). It exhibits high affinity for the Gcn5 enzyme, comparable to its natural substrate, histone H3. It shows weak inhibition on CBP (IC50=0.5 mM) and can be used in studies of cancer, metabolic, autoimmune, and neurological diseases. This product is for research use only.

  • Specific small-molecule inhibitor of histone acetyltransferase Gcn5 (IC50=100 μM).
  • High affinity for the Gcn5 enzyme, comparable to histone H3.
  • Weak inhibition on CBP (IC50=0.5 mM).
  • Used in studies of cancer, metabolic, autoimmune, and neurological diseases.
  • Purity of 99.58%.
  • Molecular weight of 184.19.
  • Chemical formula: C9H12O4.
  • Solid, white to off-white appearance.
  • Recommended storage for powder: -20°C for 3 years.
  • Solutions are unstable; freshly prepared solutions are recommended.

Catalog No. 50-003-40480 Supplier Medchemexpress LLC Supplier No. HY1290391MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyrolactone 3 (MB-3) is a specific small-molecule inhibitor of the histone acetyltransferase Gcn5 (IC50=100 μM). It exhibits high affinity for the Gcn5 enzyme, comparable to its natural substrate, histone H3. It shows weak inhibition on CBP (IC50=0.5 mM) and can be used in studies of cancer, metabolic, autoimmune, and neurological diseases. This product is for research use only.

  • Specific small-molecule inhibitor of histone acetyltransferase Gcn5 (IC50=100 μM).
  • High affinity for the Gcn5 enzyme, comparable to histone H3.
  • Weak inhibition on CBP (IC50=0.5 mM).
  • Used in studies of cancer, metabolic, autoimmune, and neurological diseases.
  • Purity of 99.58%.
  • Molecular weight of 184.19.
  • Chemical formula: C9H12O4.
  • Solid, white to off-white appearance.
  • Recommended storage for powder: -20°C for 3 years.
  • Solutions are unstable; freshly prepared solutions are recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.