Learn More
Medchemexpress LLC Butyrolactone 3 (MB-3) | 778649-18-6 | 99.6% | 10 MG
SDP
Catalog No. 5000341243
Encompass_Preferred
Butyrolactone 3 (MB-3) is a specific small-molecule inhibitor of the histone acetyltransferase Gcn5, with an IC50 of 100 μM. It demonstrates high affinity for the Gcn5 enzyme, similar to its natural substrate, histone H3, and shows weak inhibition of CBP (IC50 = 0.5 mM). This compound is suitable for studies related to cancer, metabolic, autoimmune, and neurological diseases.
  • Specific small-molecule inhibitor of histone acetyltransferase Gcn5
  • High affinity for the Gcn5 enzyme
  • Weak inhibition of CBP
  • Can be used in studies of cancer, metabolic, autoimmune, and neurological diseases
Catalog No. 50-003-41243 Supplier Medchemexpress LLC Supplier No. HY12903910MG
Butyrolactone 3 (MB-3) is a specific small-molecule inhibitor of the histone acetyltransferase Gcn5, with an IC50 of 100 μM. It demonstrates high affinity for the Gcn5 enzyme, similar to its natural substrate, histone H3, and shows weak inhibition of CBP (IC50 = 0.5 mM). This compound is suitable for studies related to cancer, metabolic, autoimmune, and neurological diseases.
  • Specific small-molecule inhibitor of histone acetyltransferase Gcn5
  • High affinity for the Gcn5 enzyme
  • Weak inhibition of CBP
  • Can be used in studies of cancer, metabolic, autoimmune, and neurological diseases