Learn More
Abcam Butyrolactone I, ATP-competitive cyclin-dependent kinase (CDK) inhibitor, 200UG
Catalog No. 502706531
Encompass_Preferred
MW 424.4 Da, Purity >97%. Cell permeable, selective, ATP-competitive CDK inhibitor. Inhibits CDC2, CDK2, and CDK5. Inhibits the cell cycle at G1/S and G2/M. Apoptotic and antitumor activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6531 Supplier Abcam Supplier No. AB141520200UG
MW 424.4 Da, Purity >97%. Cell permeable, selective, ATP-competitive CDK inhibitor. Inhibits CDC2, CDK2, and CDK5. Inhibits the cell cycle at G1/S and G2/M. Apoptotic and antitumor activity.
The product is subject to the following: Abcam Restricted Use Statement