Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyrolactone I (olomoucin) | 87414-49-1 | MFCD03453074 | 99.8% | 424.44 | C24H24O7 | 5 MG
SDP

Catalog No. 5000263275
Encompass_Preferred

Butyrolactone I (olomoucin) is an orally active, ATP-competitive cyclin-dependent kinase 1 (CDK1) inhibitor used in preclinical research to study cell-cycle regulation, apoptosis, and inflammatory signaling. It has reported activity against NF-κB and PERK/CHOP pathways and is commonly used in cancer and metabolic disease research.

  • ATP-competitive CDK1 inhibitor
  • Reported modulation of NF-κB, PERK/CHOP, and apoptosis pathways
  • High purity suitable for research applications
  • Molecular formula C24H24O7; molecular weight 424.44
  • Soluble in DMSO; ultrasonic assistance recommended
  • Store protected from light; follow supplier solvent storage guidance

Catalog No. 50-002-63275 Supplier Medchemexpress LLC Supplier No. HY1112375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyrolactone I (olomoucin) is an orally active, ATP-competitive cyclin-dependent kinase 1 (CDK1) inhibitor used in preclinical research to study cell-cycle regulation, apoptosis, and inflammatory signaling. It has reported activity against NF-κB and PERK/CHOP pathways and is commonly used in cancer and metabolic disease research.

  • ATP-competitive CDK1 inhibitor
  • Reported modulation of NF-κB, PERK/CHOP, and apoptosis pathways
  • High purity suitable for research applications
  • Molecular formula C24H24O7; molecular weight 424.44
  • Soluble in DMSO; ultrasonic assistance recommended
  • Store protected from light; follow supplier solvent storage guidance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.