Learn More
AdipoGen Butyrolactone II
SDP
Catalog No. 502562538
Encompass_Preferred
Chemical. CAS 87414-44-6. Formula C19H16O7. MW 356.3. Isolated from Aspergillus sp. Antibiotic. Weak cytotoxic towards bacterial and cancer cell lines. 5-Lipoxygenase 5-LOX inhibitor. Shows DPPH 2,2-diphenyl-1-picrylhydrazyl radical-scavening activity.
Catalog No. 50-256-2538 Supplier AdipoGen Supplier No. AGCN20423C500
Chemical. CAS 87414-44-6. Formula C19H16O7. MW 356.3. Isolated from Aspergillus sp. Antibiotic. Weak cytotoxic towards bacterial and cancer cell lines. 5-Lipoxygenase 5-LOX inhibitor. Shows DPPH 2,2-diphenyl-1-picrylhydrazyl radical-scavening activity.