Learn More
Apexbio Technology LLC BUTYRYL-CARNITINE-D3-HCL-5MG
SDP
Catalog No. 5000570380
Encompass_Preferred
Butyryl carnitine-d3 HCl (CAS No 1334532-21-6) is a deuterated compound utilized in biomedical research primarily within metabolic studies and pharmacokinetic profiling Originating as an analog of butyryl carnitine it targets carnitine transport pathways playing a role in lipid metabolism and mitochondrial function Its activity is typically observed in the nanomolar to low micromolar concentration range relevant in biochemical assays Common applications include cell models and animal studies to evaluate metabolic processes and potential drug interactions Butyryl carnitine-d3 HCl is also used in drug screening particularly for compounds affecting mitochondrial function or lipid metabolism Experimental concentrations usually vary depending on the specific research design allowing flexibility in assay development to explore therapeutic potential or mechanistic insights in various biological systems
Catalog No. 50-005-70380 Supplier Apexbio Technology LLC Supplier No. SL10295
Butyryl carnitine-d3 HCl (CAS No 1334532-21-6) is a deuterated compound utilized in biomedical research primarily within metabolic studies and pharmacokinetic profiling Originating as an analog of butyryl carnitine it targets carnitine transport pathways playing a role in lipid metabolism and mitochondrial function Its activity is typically observed in the nanomolar to low micromolar concentration range relevant in biochemical assays Common applications include cell models and animal studies to evaluate metabolic processes and potential drug interactions Butyryl carnitine-d3 HCl is also used in drug screening particularly for compounds affecting mitochondrial function or lipid metabolism Experimental concentrations usually vary depending on the specific research design allowing flexibility in assay development to explore therapeutic potential or mechanistic insights in various biological systems