Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyryl-coenzyme A sodium | 125527-24-4 | 99.0% | 859.61 | 1 MG
SDP

Catalog No. 5000349437
Encompass_Preferred

Butyryl-coenzyme A (Butyryl CoA) sodium is a coenzyme A-containing derivative of butyric acid. It is responsible for the final step of butyrate production in bacteria.

  • Coenzyme A-containing derivative of butyric acid.
  • Involved in bacterial butyrate production.
  • Available in various quantities.
  • Provided with a data sheet, COA, SDS, and handling instructions.
  • Recommended storage at -20°C, sealed, away from moisture.
  • Soluble in H2O at concentrations ≥ 100 mg/mL (116.33 mM).

Catalog No. 50-003-49437 Supplier Medchemexpress LLC Supplier No. HY1381771MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyryl-coenzyme A (Butyryl CoA) sodium is a coenzyme A-containing derivative of butyric acid. It is responsible for the final step of butyrate production in bacteria.

  • Coenzyme A-containing derivative of butyric acid.
  • Involved in bacterial butyrate production.
  • Available in various quantities.
  • Provided with a data sheet, COA, SDS, and handling instructions.
  • Recommended storage at -20°C, sealed, away from moisture.
  • Soluble in H2O at concentrations ≥ 100 mg/mL (116.33 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.