Learn More
Medchemexpress LLC Butyryl-coenzyme A (sodium) | 125527-24-4 | 99.0% | 859.61 g·mol⁻¹ | C25H41N7NaO17P3S | 5 MG
SDP
Catalog No. 5000221927
Encompass_Preferred
Butyryl-Coenzyme A (sodium) is a high-purity coenzyme A-containing derivative of butyric acid used in biochemical research. It participates in the final step of butyrate production in bacteria and is supplied as a water-soluble, white to off-white solid suitable for enzymatic and metabolic studies.
  • High purity (99.0%) for reproducible results.
  • Water soluble (≥100 mg/mL) for easy preparation and dilution.
  • White to off-white solid for consistent handling and weighing.
  • Stable under recommended storage conditions for reliable short-term and longer-term use.
  • Suitable for enzymatic, metabolic, and bacterial pathway studies.
Catalog No. 50-002-21927 Supplier Medchemexpress LLC Supplier No. HY1381775MG
Butyryl-Coenzyme A (sodium) is a high-purity coenzyme A-containing derivative of butyric acid used in biochemical research. It participates in the final step of butyrate production in bacteria and is supplied as a water-soluble, white to off-white solid suitable for enzymatic and metabolic studies.
  • High purity (99.0%) for reproducible results.
  • Water soluble (≥100 mg/mL) for easy preparation and dilution.
  • White to off-white solid for consistent handling and weighing.
  • Stable under recommended storage conditions for reliable short-term and longer-term use.
  • Suitable for enzymatic, metabolic, and bacterial pathway studies.