Learn More
Medchemexpress LLC Butyryl-L-carnitine 10mg | 162067-50-7 | 267.75 g/mol | C11H22ClNO4 | 10 MG
SDP
Catalog No. 5000229370
Encompass_Preferred
Butyryl-L-carnitine chloride is the chloride salt of an acylcarnitine that can be isolated from plasma/serum. It is supplied as a high-purity solid suitable for analytical and biochemical research applications.
  • High purity (≈98.1%) for reliable analytical results.
  • Provided as a solid for easy handling and storage.
  • CAS number 162067-50-7 for unambiguous identification.
  • Compatible with biochemical and mass spectrometry assays.
  • Available in small pack sizes for research-scale use.
Catalog No. 50-002-29370 Supplier Medchemexpress LLC Supplier No. HY11570410MG
Butyryl-L-carnitine chloride is the chloride salt of an acylcarnitine that can be isolated from plasma/serum. It is supplied as a high-purity solid suitable for analytical and biochemical research applications.
  • High purity (≈98.1%) for reliable analytical results.
  • Provided as a solid for easy handling and storage.
  • CAS number 162067-50-7 for unambiguous identification.
  • Compatible with biochemical and mass spectrometry assays.
  • Available in small pack sizes for research-scale use.