Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyryl-L-carnitine- 1mg | 1334532-21-6 | 270.77 | C11H19D3ClNO4 | 1 MG
SDP

Catalog No. 5000307697
Encompass_Preferred

Butyryl-L-carnitine-d3 chloride is a deuterium-labeled analogue of butyryl-L-carnitine intended for use as an isotopic tracer and internal standard in mass spectrometry and metabolic tracing applications. Supplied as a high-purity stable isotope, it enables accurate quantitation and reliable identification of butyrylcarnitine in biological samples.

  • Deuterium-labeled (d3) analogue for use as an isotopic tracer.
  • High purity: 99.91% as supplied.
  • Molecular weight 270.77; formula C11H19D3ClNO4.
  • Suitable as an internal standard for LC-MS and GC-MS quantitation.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store sealed away from moisture; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-003-07697 Supplier Medchemexpress LLC Supplier No. HY115704S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyryl-L-carnitine-d3 chloride is a deuterium-labeled analogue of butyryl-L-carnitine intended for use as an isotopic tracer and internal standard in mass spectrometry and metabolic tracing applications. Supplied as a high-purity stable isotope, it enables accurate quantitation and reliable identification of butyrylcarnitine in biological samples.

  • Deuterium-labeled (d3) analogue for use as an isotopic tracer.
  • High purity: 99.91% as supplied.
  • Molecular weight 270.77; formula C11H19D3ClNO4.
  • Suitable as an internal standard for LC-MS and GC-MS quantitation.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store sealed away from moisture; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.