Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyryl-L-carnitine 25mg | 162067-50-7 | 25 MG
SDP

Catalog No. 5000226518
Encompass_Preferred

Butyryl-L-carnitine chloride is an acylcarnitine research reagent used as a reference standard in metabolic, analytical, and pharmacological studies. It is supplied as a white to off-white solid with high reported purity.

  • White to off-white solid, easily handled in laboratory settings.
  • Reported purity 98.1% (manufacturer listing).
  • CAS number 162067-50-7 for unambiguous identification.
  • Chemical formula C11H22ClNO4 and molecular weight 267.75 g/mol.
  • Suitable as a reference standard for metabolic and analytical assays.
  • Available in small research sizes for analytical use.

Catalog No. 50-002-26518 Supplier Medchemexpress LLC Supplier No. HY11570425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyryl-L-carnitine chloride is an acylcarnitine research reagent used as a reference standard in metabolic, analytical, and pharmacological studies. It is supplied as a white to off-white solid with high reported purity.

  • White to off-white solid, easily handled in laboratory settings.
  • Reported purity 98.1% (manufacturer listing).
  • CAS number 162067-50-7 for unambiguous identification.
  • Chemical formula C11H22ClNO4 and molecular weight 267.75 g/mol.
  • Suitable as a reference standard for metabolic and analytical assays.
  • Available in small research sizes for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.