Learn More
Medchemexpress LLC Butyryl-L-carnitine- 5mg | 1334532-21-6 | 270.77 g/mol | C11H19D3ClNO4 | 5 MG
SDP
Catalog No. 5000227944
Encompass_Preferred
Butyryl-L-carnitine-d3 chloride is the deuterium-labeled form of butyryl-L-carnitine (chloride) intended for use as a tracer or internal standard in quantitative analytical methods. It is supplied in small milligram quantities with high isotopic and chemical purity for laboratory research and method development.
Catalog No. 50-002-27944 Supplier Medchemexpress LLC Supplier No. HY115704S15MG
Butyryl-L-carnitine-d3 chloride is the deuterium-labeled form of butyryl-L-carnitine (chloride) intended for use as a tracer or internal standard in quantitative analytical methods. It is supplied in small milligram quantities with high isotopic and chemical purity for laboratory research and method development.