Learn More
Medchemexpress LLC Butyryl-L-carnitine 5mg | 162067-50-7 | 267.75 g/mol | C11H22ClNO4 | 5 MG
SDP
Catalog No. 5000226302
Encompass_Preferred
Butyryl-L-carnitine chloride is the butyryl ester of L-carnitine and an acylcarnitine that can be isolated from plasma or serum. It is supplied as a purified research reagent for biochemical and pharmacological studies of fatty acid metabolism and transporter activity. The chloride salt has formula C11H22ClNO4, molecular weight 267.75 g/mol, and reported purity around 98%.
  • Purity 98.07%.
  • Solid form suitable for storage and handling.
  • Used for studying fatty acid metabolism and acylcarnitine profiling.
  • Appropriate for analytical and biochemical assays.
Catalog No. 50-002-26302 Supplier Medchemexpress LLC Supplier No. HY1157045MG
Butyryl-L-carnitine chloride is the butyryl ester of L-carnitine and an acylcarnitine that can be isolated from plasma or serum. It is supplied as a purified research reagent for biochemical and pharmacological studies of fatty acid metabolism and transporter activity. The chloride salt has formula C11H22ClNO4, molecular weight 267.75 g/mol, and reported purity around 98%.
  • Purity 98.07%.
  • Solid form suitable for storage and handling.
  • Used for studying fatty acid metabolism and acylcarnitine profiling.
  • Appropriate for analytical and biochemical assays.