Learn More
Medchemexpress LLC Butyrylcarnitine | 25576-40-3 | 100.0% | 231.29 | 100 MG
SDP
Catalog No. 5000437237
Encompass_Preferred
Butyrylcarnitine is an endogenous metabolite found in plasma. Elevated levels are associated with abnormalities in lipid and energy metabolism. It can serve as a diagnostic and prognostic indicator for diseases like heart failure and head and neck cancer.
  • Endogenous metabolite
  • Associated with lipid and energy metabolism
  • Diagnostic and prognostic indicator for certain diseases
  • Promotes cell proliferation in specific cell lines
  • Enhances proliferation, migration, and invasion abilities in HN30 cells
Catalog No. 50-004-37237 Supplier Medchemexpress LLC Supplier No. HY113168100MG
Butyrylcarnitine is an endogenous metabolite found in plasma. Elevated levels are associated with abnormalities in lipid and energy metabolism. It can serve as a diagnostic and prognostic indicator for diseases like heart failure and head and neck cancer.
  • Endogenous metabolite
  • Associated with lipid and energy metabolism
  • Diagnostic and prognostic indicator for certain diseases
  • Promotes cell proliferation in specific cell lines
  • Enhances proliferation, migration, and invasion abilities in HN30 cells