Learn More
Medchemexpress LLC Butyrylcarnitine | 25576-40-3 | 100.0% | 231.29 | 25 MG
SDP
Catalog No. 5000439572
Encompass_Preferred
Butyrylcarnitine is an endogenous metabolite found in plasma. Elevated levels are associated with abnormalities in lipid and energy metabolism. It can serve as a diagnostic and prognostic indicator for certain diseases, including heart failure and head and neck cancer.
  • Endogenous metabolite.
  • Associated with lipid and energy metabolism.
  • Potential diagnostic and prognostic indicator for heart failure and head and neck cancer.
Catalog No. 50-004-39572 Supplier Medchemexpress LLC Supplier No. 25MGHY113168
Butyrylcarnitine is an endogenous metabolite found in plasma. Elevated levels are associated with abnormalities in lipid and energy metabolism. It can serve as a diagnostic and prognostic indicator for certain diseases, including heart failure and head and neck cancer.
  • Endogenous metabolite.
  • Associated with lipid and energy metabolism.
  • Potential diagnostic and prognostic indicator for heart failure and head and neck cancer.