Learn More
TARGETMOL CHEMICALS INC Butyrylcarnitine 25MG
SDP
Catalog No. 502878536
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Butyrylcarnitine is a plasma metabolite that is an indicator of abnormal metabolism due to disease and is used in the diagnosis of heart failure and pancreatic cysts. Purity 99%
Catalog No. 50-287-8536 Supplier TARGETMOL CHEMICALS INC Supplier No. T1484025MG
Also available in 2 mg, 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Butyrylcarnitine is a plasma metabolite that is an indicator of abnormal metabolism due to disease and is used in the diagnosis of heart failure and pancreatic cysts. Purity 99%