Learn More
Medchemexpress LLC Butyrylcholine chloride | 2963-78-2 | MFCD00011844 | 98.0% | 209.71 g/mol | C9H20ClNO2 | 100 MG
SDP
Catalog No. 5000303302
Encompass_Preferred
Butyrylcholine chloride is a choline ester used as a substrate in enzymatic and biochemical assays to measure butyrylcholinesterase and acetylcholinesterase activity. It is supplied as a solid with high reported purity and supporting analytical documentation for laboratory use.
  • Substrate for BChE and AChE activity assays.
  • High purity (98.0%).
  • Solid form suitable for storage and precise weighing.
  • Molecular weight 209.71 g/mol.
  • Analytical documentation available (SDS, COA, datasheet).
Catalog No. 50-003-03302 Supplier Medchemexpress LLC Supplier No. HYW014419100MG
Butyrylcholine chloride is a choline ester used as a substrate in enzymatic and biochemical assays to measure butyrylcholinesterase and acetylcholinesterase activity. It is supplied as a solid with high reported purity and supporting analytical documentation for laboratory use.
  • Substrate for BChE and AChE activity assays.
  • High purity (98.0%).
  • Solid form suitable for storage and precise weighing.
  • Molecular weight 209.71 g/mol.
  • Analytical documentation available (SDS, COA, datasheet).