Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyrylcholine chloride | 2963-78-2 | MFCD00011844 | 98.0% | 209.71 g/mol | C9H20ClNO2 | 250 MG
SDP

Catalog No. 5000302483
Encompass_Preferred

Butyrylcholine chloride is a choline ester supplied as a solid chloride salt for use as a biochemical assay reagent and substrate in cholinesterase and enzymatic activity assays. It is provided at high purity for research applications and is accompanied by product documentation for handling and characterization.

  • High purity (98.0%) suitable for enzymatic assays.
  • Solid chloride salt, stable for sealed storage.
  • Molecular weight 209.71 g/mol; formula C9H20ClNO2.
  • Available in small research package sizes, including 250 mg.
  • Documentation available: data sheet, SDS, COA.
  • Store sealed, away from moisture at -20°C; in solution store at -80°C up to 6 months.

Catalog No. 50-003-02483 Supplier Medchemexpress LLC Supplier No. 250MGHYW014419
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyrylcholine chloride is a choline ester supplied as a solid chloride salt for use as a biochemical assay reagent and substrate in cholinesterase and enzymatic activity assays. It is provided at high purity for research applications and is accompanied by product documentation for handling and characterization.

  • High purity (98.0%) suitable for enzymatic assays.
  • Solid chloride salt, stable for sealed storage.
  • Molecular weight 209.71 g/mol; formula C9H20ClNO2.
  • Available in small research package sizes, including 250 mg.
  • Documentation available: data sheet, SDS, COA.
  • Store sealed, away from moisture at -20°C; in solution store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.