Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyrylcholine iodide | 2494-56-6 | 99.8% | 301.17 | 250 MG
SDP

Catalog No. 5000366407
Encompass_Preferred

Butyrylcholine iodide is a biochemical reagent intended for life science related research. It serves as a biological material or organic compound for various research applications.

  • For research use only
  • Not sold to patients
  • Molecular weight: 301.17
  • Purity: 99.83%

Catalog No. 50-003-66407 Supplier Medchemexpress LLC Supplier No. 250MGHYW009236
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyrylcholine iodide is a biochemical reagent intended for life science related research. It serves as a biological material or organic compound for various research applications.

  • For research use only
  • Not sold to patients
  • Molecular weight: 301.17
  • Purity: 99.83%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.