Learn More
Cayman Chemical ButyrylCoenzyme AsodIM sa 10mg
SDP
Catalog No. 502937425
Encompass_Preferred
A short-chain acyl CoA; an intermediate in the synthesis of butyrate
Catalog No. 50-293-7425 Supplier Cayman Chemical Supplier No. 2786510 MG
A short-chain acyl CoA; an intermediate in the synthesis of butyrate