Learn More
Cayman Chemical Butyryl-L-carnitine (chloride), Purity: ≥95%, CAS: 162067-50-7, 100 mg
SDP
Catalog No. 502934650
Encompass_Preferred
A butyrate ester of carnitine; blocks carnitine uptake by the carnitine transporter and glycine transport by the amino acid transporter in HRPE cells (IC50s = 1.5 µM and 4.6 mM, respectively)
Catalog No. 50-293-4650 Supplier Cayman Chemical Supplier No. 26542100 MG
A butyrate ester of carnitine; blocks carnitine uptake by the carnitine transporter and glycine transport by the amino acid transporter in HRPE cells (IC50s = 1.5 µM and 4.6 mM, respectively)