Learn More
Cayman Chemical ButyrylLcarnItInchLRIde 50mg
SDP
Catalog No. 502934652
Encompass_Preferred
A butyrate ester of carnitine; blocks carnitine uptake by the carnitine transporter and glycine transport by the amino acid transporter in HRPE cells (IC50s = 1.5 µM and 4.6 mM, respectively)
Catalog No. 50-293-4652 Supplier Cayman Chemical Supplier No. 2654250 MG
A butyrate ester of carnitine; blocks carnitine uptake by the carnitine transporter and glycine transport by the amino acid transporter in HRPE cells (IC50s = 1.5 µM and 4.6 mM, respectively)