Learn More
AdipoGen Butyryloxy-pyren-trisulfonic
SDP
Supplier:  AdipoGen CDXB0025M050
Encompass_Preferred
Chemical. CAS 115787-82-1. Formula C20H13Na3O11S3. MW 594.48. Synthetic. Highly water soluble enzyme substrate for the fluorimetric assay of esterases at longwave excitation and emission wavelengths.
Catalog No. 50-256-1705