Learn More
Medchemexpress LLC Butyryltimolol | 106351-79-5 | 98.0% | 386.51 g·mol⁻¹ | C17H30N4O4S | 10 MG
SDP
Catalog No. 5000247088
Encompass_Preferred
Butyryltimolol is a butyryl ester prodrug of timolol that enhances corneal penetration and acts as a β-adrenergic blocker for ocular research applications, including studies of drug delivery and glaucoma pharmacology.
  • Improves corneal penetration compared with timolol
  • Functions as a β-adrenergic blocker
  • High purity (98.0%) suitable for research
  • Liquid form, colorless to light yellow
  • Recommended storage: pure form -20°C (3 years), in solvent -80°C (6 months)
Catalog No. 50-002-47088 Supplier Medchemexpress LLC Supplier No. HY10203210MG
Butyryltimolol is a butyryl ester prodrug of timolol that enhances corneal penetration and acts as a β-adrenergic blocker for ocular research applications, including studies of drug delivery and glaucoma pharmacology.
  • Improves corneal penetration compared with timolol
  • Functions as a β-adrenergic blocker
  • High purity (98.0%) suitable for research
  • Liquid form, colorless to light yellow
  • Recommended storage: pure form -20°C (3 years), in solvent -80°C (6 months)