Learn More
Medchemexpress LLC Butyryltimolol | 106351-79-5 | 98.0% | 386.51 g/mol | C17H30N4O4S | 100 MG
SDP
Catalog No. 5000247838
Encompass_Preferred
Butyryltimolol is a lipophilic ester prodrug of timolol that enhances corneal penetration and functions as a β-adrenergic blocker. Supplied for research use, it is used to evaluate improved ocular delivery and β-adrenergic receptor activity in preclinical studies.
  • Improves corneal penetration of timolol
  • Acts as a β-adrenergic blocker for receptor studies
  • Lipophilic ester prodrug to enhance ocular absorption
  • High purity suitable for research applications
  • Stable when stored under recommended conditions
Catalog No. 50-002-47838 Supplier Medchemexpress LLC Supplier No. HY102032100MG
Butyryltimolol is a lipophilic ester prodrug of timolol that enhances corneal penetration and functions as a β-adrenergic blocker. Supplied for research use, it is used to evaluate improved ocular delivery and β-adrenergic receptor activity in preclinical studies.
  • Improves corneal penetration of timolol
  • Acts as a β-adrenergic blocker for receptor studies
  • Lipophilic ester prodrug to enhance ocular absorption
  • High purity suitable for research applications
  • Stable when stored under recommended conditions