Learn More
Medchemexpress LLC Butyzamide | 1110767-45-7 | 99.8% | 591.55 | 1 MG
SDP
Catalog No. 5000362623
Encompass_Preferred
Butyzamide is an orally active activator of Mpl, a thrombopoietin (TPO) receptor. It increases the phosphorylation level of JAK2, STAT3, STAT5, and MAPK, and has been shown to increase platelet levels in mouse xenotransplantation assays.
  • Increases phosphorylation of JAK2, STAT3, STAT5, and MAPK.
  • Raises platelet levels in mouse xenotransplantation assays.
  • Induces colony-forming unit-megakaryocyte and polyploid megakaryocytes.
  • Orally active Mpl activator.
Catalog No. 50-003-62623 Supplier Medchemexpress LLC Supplier No. HY1487481MG
Butyzamide is an orally active activator of Mpl, a thrombopoietin (TPO) receptor. It increases the phosphorylation level of JAK2, STAT3, STAT5, and MAPK, and has been shown to increase platelet levels in mouse xenotransplantation assays.
  • Increases phosphorylation of JAK2, STAT3, STAT5, and MAPK.
  • Raises platelet levels in mouse xenotransplantation assays.
  • Induces colony-forming unit-megakaryocyte and polyploid megakaryocytes.
  • Orally active Mpl activator.