Learn More
BD Cell Analysis 3P BUV496 Rat Anti-Mouse CD44, 50 ug
Supplier:  BD Cell Analysis 3P 741057
Encompass
This antibody was developed for use in flow cytometry. The production process underwent stringent testing and validation to assure that it generates a high-quality conjugate with consistent performance and specific binding activity. However, verification testing has not been performed on all conjugate lots.
Catalog No. NC1712598
Please call customer service for current pricing. 1-800-766-7000