Learn More
BD Cell Analysis 3P BUV805 Rat Anti-Mouse Ly-6G, 50 µg
Supplier:  BD Cell Analysis 3P 741994
Encompass
Concentration: 0.2 mg/ml, Isotype: Rat LEW, also known as Lewis IgG2a, κ; Reactivity: Mouse (Tested in Development), Application: Flow cytometry (Qualified), Immunogen: Ly-6G-transfected EL4J Cell Line, Alternative Name: Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1
Catalog No. NC1648710