Learn More
Sigma Aldrich Fine Chemicals Biosciences PI 3-Kinase HTRF(R) Assay; 1920 wells The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 5 plates. | 1ML
Catalog No. NC1869245
Encompass
PI 3-Kinase HTRF(R) Assay; 1920 wells The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 5 plates. | 1ML
Catalog No. NC1869245 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 33017
PI 3-Kinase HTRF(R) Assay; 1920 wells The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 5 plates. | 1ML