Learn More
MSC Buyers Products Rubber Wheel Chock, 8 in Overall Length, Smooth Base Tread
Catalog No. 5000514165
Encompass
This wheel chock is designed to secure vehicles and prevent accidental movement. Constructed from durable molded rubber, it features an 8-inch overall length and a smooth base tread for reliable grip, complemented by a ridged surface tread for enhanced tire traction. It also includes a chain eye bolt for convenient tethering.
  • Molded rubber construction ensures durability.
  • Smooth base tread provides stable positioning.
  • Ridged surface tread enhances tire grip.
  • Includes a chain eye bolt for easy tethering.
Catalog No. 50-005-14165 Supplier MSC Supplier No. 38134169
This wheel chock is designed to secure vehicles and prevent accidental movement. Constructed from durable molded rubber, it features an 8-inch overall length and a smooth base tread for reliable grip, complemented by a ridged surface tread for enhanced tire traction. It also includes a chain eye bolt for convenient tethering.
  • Molded rubber construction ensures durability.
  • Smooth base tread provides stable positioning.
  • Ridged surface tread enhances tire grip.
  • Includes a chain eye bolt for easy tethering.