Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pain Care Labs Buzzy™ Mini Healthcare
SDP

Catalog No. 22450105
Change view
Click to view available options
Color:
LadyBuzz Red
Plain Black
Yellow and Black Striped
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-450-105 LadyBuzz Red
22-450-104 Plain Black
22-450-103 Yellow and Black Striped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-450-105 Supplier Pain Care Labs Supplier No. BKH M3
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pain Care Labs Buzzy™ Mini Healthcare features a contoured and compact design which tucks under a standard tourniquet, or fits easily in the hand to deliver powerful pain blocking right where you need it.

  • Latex-free
  • Intended for healthcare and multi-patient use.
  • Features 3 minute auto-shut off and 2 vibration settings.
  • Includes 4 Universal Healthcare Wings, 1 Silicone Comfort Strap, 2 AAA Batteries (preinstalled in unit), 1 Instruction Manual
  • Reusable medical device that should be thoroughly cleaned and reprocessed following your facility's infection control protocol for non-critical equipment.

Specifications

Product Type Medical Device
Color LadyBuzz Red
For Use With (Application) Needle procedures
Material Medical Grade Plastic
Size Mini
Dimensions (L x W x H) 4.75 x 2.5 x 7.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.