Learn More
Pain Care Labs Buzzy™ Universal Healthcare Ice Wings

Description
- Latex-free
- Intended for multi-patient use.
- Simply clean per your facility's infection control policy states for non-critical, reusable equipment such as beds and monitors.
- Stays frozen for approximately 10 minutes.
- Wipe clean, refreeze and reuse.
Specifications
Specifications
| Quantity | 25 Pk. |
| Product Type | Cold Pack |
| Color | Blue |
| For Use With (Application) | Needle Procedures |
| Material | Polyethylene |
| Size | Universal |
| Dimensions (L x W x H) | 18 x 8 x 26 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.