Learn More
Pain Care Labs Buzzy™ Universal Soft Ice Wings

Description
- Latex-free
- Excellent disposable option
- Not for multi-patient use
- Work with Buzzy™ Mini (attach to the clip), or Buzzy™ XL Models (just slide the wings underneath the elastic strap)
- Reversible gel wings have a colder blue side and a less intense soft white side
Specifications
Specifications
| Quantity | 100 Pk. |
| Product Type | Cold Pack |
| Color | Blue and White |
| For Use With (Application) | Needle Procedures |
| Material | Polyethylene |
| Size | Universal |
| Dimensions (L x W x H) | 18 x 8 x 26 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.