Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pain Care Labs Buzzy™ XL Healthcare
SDP

Catalog No. 22450102
Change view
Click to view available options
Color:
LadyBuzz Red
Yellow and Black Striped
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-450-102 LadyBuzz Red
22-450-100 Yellow and Black Striped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-450-102 Supplier Pain Care Labs Supplier No. BTH 3
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pain Care Labs Buzzy™ XL Healthcare is proven in clinical trials to decrease the pain of IV insertion, and lets patients know pain relief is a priority.

  • Latex-free
  • Intended for healthcare and multi-patient use.
  • Includes 4 Universal Healthcare Wings, 4 Silicone Comfort Strap, 2 AAA Batteries (preinstalled in unit), 1 Instruction Manual
  • Reusable medical device that should be thoroughly cleaned and reprocessed following your facility’s infection control protocol for non-critical equipment.
  • Stand up to hospital grade cleansers.
  • Use with disposable hospital tourniquets or press in place for pain relief on contact.

Specifications

Product Type Medical Device
Color LadyBuzz Red
For Use With (Application) Needle Procedures
Material Medical Grade Plastic
Size X-Large
Dimensions (L x W x H) 5.5 x 3 x 8.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.