Learn More
Jackson Immuno Research Labs Brilliant Violet 421™ AffiniPure Goat Anti-Mouse IgG, Fcγ fragment specific | 0.25mL
SDP
Supplier:  Jackson Immuno Research Labs 115675071
Encompass_Preferred
Target: Mouse | Host: Goat | Antibody Format: Whole IgG | Specificity: IgG, Fcγ fragment specific | Minimal Cross Reactivity: Human, Bovine, Horse Serum Proteins | Conjugate: Brilliant Violet 421™
Catalog No. NC1320740
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov