Learn More
Biolegend Brilliant Violet 510™ anti-human CD206 (MMR)
Supplier:  Biolegend 321138
Encompass_Preferred
Brilliant Violet 510™ anti-human CD206 (MMR) [15-2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1488761