Learn More
Biolegend Brilliant Violet 605™ anti-mouse NK-1.1
Supplier:  Biolegend 108753
Encompass_Preferred
Brilliant Violet 605™ anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1429849