Learn More
Biolegend Brilliant Violet 650™ anti-mouse CD62L
Supplier:  Biolegend 104453
Encompass_Preferred
Brilliant Violet 650™ anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1906611