Learn More
Biolegend Brilliant Violet 711™ anti-human TCR Vδ2
Supplier:  Biolegend 331412
Encompass_Preferred
Brilliant Violet 711™ anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactive: Chimpanzee, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1838382