Learn More
Biolegend Brilliant Violet 785™ anti-human CD123
Supplier:  Biolegend 306032
Encompass_Preferred
Brilliant Violet 785™ anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1234804