Learn More
Biolegend Brilliant Violet 711™ anti-mouse CD366 (Tim-3)
Supplier:  Biolegend 119727
Encompass_Preferred
Brilliant Violet 711™ anti-mouse CD366 (Tim-3) [RMT3-23]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1424748