Learn More
Medchemexpress LLC BV02 | 292870-53-2 | MFCD00356631 | 99.8% | 377.36 | C20H15N3O5 | 50 MG
SDP
Catalog No. 5000249178
Encompass_Preferred
BV02 is a small-molecule research compound that inhibits 14-3-3 protein-protein interactions and has reported antibacterial activity. It modulates cell signaling by enhancing PARP and caspase-3 cleavage, and by inducing apoptosis and autophagy, making it useful for cellular and infection-related studies.
  • Inhibits 14-3-3 protein-protein interactions.
  • Induces apoptosis and autophagy in cell-based assays.
  • Enhances PARP and caspase-3 cleavage.
  • Shows reported antibacterial activity.
  • Supplied at high purity suitable for research use.
  • Available in solid and ready-to-use solution formats.
Catalog No. 50-002-49178 Supplier Medchemexpress LLC Supplier No. HY10198550MG
BV02 is a small-molecule research compound that inhibits 14-3-3 protein-protein interactions and has reported antibacterial activity. It modulates cell signaling by enhancing PARP and caspase-3 cleavage, and by inducing apoptosis and autophagy, making it useful for cellular and infection-related studies.
  • Inhibits 14-3-3 protein-protein interactions.
  • Induces apoptosis and autophagy in cell-based assays.
  • Enhances PARP and caspase-3 cleavage.
  • Shows reported antibacterial activity.
  • Supplied at high purity suitable for research use.
  • Available in solid and ready-to-use solution formats.