Learn More
Medchemexpress LLC Bv02 | 292870-53-2 | MFCD00356631 | 99.8% | 377.36 g/mol | C20H15N3O5 | 10 MG
SDP
Catalog No. 5000249166
Encompass_Preferred
BV02 is a small-molecule inhibitor of 14-3-3 protein-protein interactions used in preclinical research to study apoptosis, autophagy, Akt signaling, and host-pathogen responses. It is provided for in vitro experiments in solid form and as a DMSO solution.
  • High chemical purity (≈99.8%).
  • Well-defined molecular formula C20H15N3O5 and molecular weight 377.36 g/mol.
  • Indexed by CAS number 292870-53-2 for unambiguous identification.
  • Available in multiple solid sizes and as a 10 mM solution in DMSO for assay readiness.
  • Reported to modulate PARP and caspase-3 cleavage and to affect Akt signaling in cell studies.
  • Supplied for research use only; not for human or veterinary use.
Catalog No. 50-002-49166 Supplier Medchemexpress LLC Supplier No. HY10198510MG
BV02 is a small-molecule inhibitor of 14-3-3 protein-protein interactions used in preclinical research to study apoptosis, autophagy, Akt signaling, and host-pathogen responses. It is provided for in vitro experiments in solid form and as a DMSO solution.
  • High chemical purity (≈99.8%).
  • Well-defined molecular formula C20H15N3O5 and molecular weight 377.36 g/mol.
  • Indexed by CAS number 292870-53-2 for unambiguous identification.
  • Available in multiple solid sizes and as a 10 mM solution in DMSO for assay readiness.
  • Reported to modulate PARP and caspase-3 cleavage and to affect Akt signaling in cell studies.
  • Supplied for research use only; not for human or veterinary use.