Learn More
Medchemexpress LLC BV02 | 292870-53-2 | MFCD00356631 | 99.8% | 377.36 g/mol | C20H15N3O5 | 5 MG
SDP
Catalog No. 5000248473
Encompass_Preferred
BV02 is a small-molecule 14-3-3 protein-protein interaction inhibitor provided as a white to off-white solid for research use. It has been reported to induce apoptosis and autophagy and to display antibacterial activity in preclinical studies. The product is supplied with stability guidelines for powder and solution storage.
Catalog No. 50-002-48473 Supplier Medchemexpress LLC Supplier No. HY1019855MG
BV02 is a small-molecule 14-3-3 protein-protein interaction inhibitor provided as a white to off-white solid for research use. It has been reported to induce apoptosis and autophagy and to display antibacterial activity in preclinical studies. The product is supplied with stability guidelines for powder and solution storage.