Learn More
Alkali Scientific BV1010 BenchMixer XL Multi-Tub
SDP
Catalog No. 503029624
Encompass_Preferred
The Alkali Scientific(BV1010) BenchMixerXL is a heavy-duty Multi Tube Vortex Mixer designed to facilitate hands-free mixing in tubes flasks vials or cylinders It is ideal for the QuEChERS pesticide analysis method and is instantly adaptable to an exceptionally broad range of vessels racks and applications (horizontal or vertical) The precisely balanced drive system includes an oversized motor that delivers accurate reproducible speeds up to 2500rpm with rattle-free operation and minimal vibration The programmable pulsing function allows for time specific interval mixing increasing agitation The 712 inch platform accommodates a wide variety of standard pre-drilled tube racks Additionally BenchMixer XL can easily be adapted to almost any type of vessel commonly found in the laboratory (contact us for details) Using the BenchMixer XL is easy after loading the sample rack and tightening the locking knobs select the desired speed and run time on the large digital control panel Then s
Catalog No. 50-302-9624 Supplier Alkali Scientific Supplier No. RS7003
The Alkali Scientific(BV1010) BenchMixerXL is a heavy-duty Multi Tube Vortex Mixer designed to facilitate hands-free mixing in tubes flasks vials or cylinders It is ideal for the QuEChERS pesticide analysis method and is instantly adaptable to an exceptionally broad range of vessels racks and applications (horizontal or vertical) The precisely balanced drive system includes an oversized motor that delivers accurate reproducible speeds up to 2500rpm with rattle-free operation and minimal vibration The programmable pulsing function allows for time specific interval mixing increasing agitation The 712 inch platform accommodates a wide variety of standard pre-drilled tube racks Additionally BenchMixer XL can easily be adapted to almost any type of vessel commonly found in the laboratory (contact us for details) Using the BenchMixer XL is easy after loading the sample rack and tightening the locking knobs select the desired speed and run time on the large digital control panel Then s