Learn More
Biolegend Brilliant Violet 421™ anti-human CD64
Supplier:  Biolegend 305019
Encompass_Preferred
Brilliant Violet 421™ anti-human CD64 [10.1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. NC1363790